Assay Details
| Gene Symbol: MFSD6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709755.3 | XP_005709812.2 | 1 | 1840 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFSD6L| Gene Symbol: MFSD6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709755.3 | XP_005709812.2 | 1 | 1840 | 64 | |
Set Membership: |
|