Assay Details
| Gene Symbol: LOC108634984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045300.1 | XP_017900789.1 | 1 - 2 | 1044 | 98 | |
| Gene Symbol: WDR60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046629.1 | XP_017902118.1 | 24 - 25 | 3232 | 98 | |
| XM_018046630.1 | XP_017902119.1 | 23 - 24 | 3166 | 98 | ||
| XM_018046631.1 | XP_017902120.1 | 21 - 22 | 3004 | 98 | ||
More Information
Set Membership: |
|

LOC108634984
Genomic Map