Assay Details
| Gene Symbol: C7H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051347.1 | XP_017906836.1 | 5 - 6 | 1648 | 91 | |
| GenBank mRNA | JO434364.2 | - | 3 - 4 | 1368 | 91 | |
| JO621810.2 | - | 3 - 4 | 799 | 91 | ||
More Information
Set Membership: |
|

C7H19orf44
Genomic Map