Assay Details
| Gene Symbol: PCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054616.1 | XP_017910105.1 | 3 - 4 | 580 | 65 | |
| GenBank mRNA | JO426972.1 | - | 3 - 4 | 644 | 65 | |
More Information
Set Membership: |
|

PCK2
Genomic Map