Assay Details
| Gene Symbol: HLCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052778.1 | XP_017908267.1 | 6 - 7 | 1946 | 55 | |
| GenBank mRNA | KA329349.1 | - | 5 - 6 | 1107 | 55 | |
More Information
Set Membership: |
|

HLCS
Genomic Map