Assay Details
| Gene Symbol: KYAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055925.1 | XP_017911414.1 | 4 - 5 | 439 | 55 | |
| XM_018055926.1 | XP_017911415.1 | 3 - 4 | 650 | 55 | ||
| XM_018055927.1 | XP_017911416.1 | 3 - 4 | 319 | 55 | ||
| XM_018055928.1 | XP_017911417.1 | 2 - 3 | 203 | 55 | ||
| XM_018055929.1 | XP_017911418.1 | 2 - 3 | 476 | 55 | ||
| XM_018055930.1 | XP_017911419.1 | 3 - 4 | 235 | 55 | ||
| XM_018055931.1 | XP_017911420.1 | 2 - 3 | 152 | 55 | ||
| XM_018055932.1 | XP_017911421.1 | 3 - 4 | 205 | 55 | ||
| XM_018055933.1 | XP_017911422.1 | 2 - 3 | 189 | 55 | ||
| GenBank mRNA | JO603663.2 | - | 1 - 2 | 209 | 55 | |
More Information
Set Membership: |
|

KYAT1
Genomic Map