Assay Details
| Gene Symbol: TSC22D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056767.1 | XP_017912256.1 | 3 | 3685 | 85 | |
| XM_018056771.1 | XP_017912260.1 | 4 | 3322 | 85 | ||
| XM_018056773.1 | XP_017912262.1 | 3 | 1037 | 85 | ||
| XM_018056776.1 | XP_017912265.1 | 3 | 681 | 85 | ||
| GenBank mRNA | JO632864.2 | - | 3 | 841 | 85 | |
More Information
Set Membership: |
|

TSC22D1
Genomic Map