Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063090.1 | XP_017918579.1 | 3 - 4 | 472 | 52 | |
| XM_018063091.1 | XP_017918580.1 | 3 - 4 | 466 | 52 | ||
| GenBank mRNA | JO641256.2 | - | 4 - 5 | 578 | 52 | |
| KA335725.1 | - | 2 - 3 | 339 | 52 | ||
More Information
Set Membership: |
|

SYT5
Genomic Map