Assay Details
| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 12 - 13 | 1921 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102171917| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 12 - 13 | 1921 | 84 | |
Set Membership: |
|