Assay Details
| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 16 - 17 | 2448 | 93 | |
| XM_018065327.1 | XP_017920816.1 | 5 - 6 | 1082 | 93 | ||
| GenBank mRNA | JT676791.1 | - | 14 - 15 | 2092 | 93 | |
More Information
Set Membership: |
|

LOC102171917
Genomic Map