Assay Details
| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 18 - 19 | 2737 | 92 | |
| XM_018065327.1 | XP_017920816.1 | 7 - 8 | 1371 | 92 | ||
More Information
Set Membership: |
|

LOC102171917
Genomic Map