Assay Details
| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 6 - 7 | 941 | 77 | |
| GenBank mRNA | JT676791.1 | - | 7 - 8 | 984 | 77 | |
More Information
Set Membership: |
|

LOC102171917
Genomic Map