Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 19 - 20 | 3054 | 66 | |
| XM_018063927.1 | XP_017919416.1 | 19 - 20 | 2705 | 66 | ||
| XM_018063928.1 | XP_017919417.1 | 19 - 20 | 2656 | 66 | ||
| XM_018063929.1 | XP_017919418.1 | 18 - 19 | 2952 | 66 | ||
| XM_018063931.1 | XP_017919420.1 | 18 - 19 | 2943 | 66 | ||
| XM_018063932.1 | XP_017919421.1 | 19 - 20 | 3054 | 66 | ||
| GenBank mRNA | KA327226.1 | - | 19 - 20 | 2768 | 66 | |
More Information
Set Membership: |
|

LOC102178109
Genomic Map