Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 26 - 27 | 4001 | 80 | |
| XM_018063927.1 | XP_017919416.1 | 26 - 27 | 3652 | 80 | ||
| XM_018063928.1 | XP_017919417.1 | 26 - 27 | 3603 | 80 | ||
| XM_018063929.1 | XP_017919418.1 | 25 - 26 | 3899 | 80 | ||
| XM_018063931.1 | XP_017919420.1 | 25 - 26 | 3890 | 80 | ||
| XM_018063932.1 | XP_017919421.1 | 26 - 27 | 4001 | 80 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map