Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 28 - 29 | 4169 | 98 | |
| XM_018063927.1 | XP_017919416.1 | 28 - 29 | 3820 | 98 | ||
| XM_018063928.1 | XP_017919417.1 | 28 - 29 | 3771 | 98 | ||
| XM_018063929.1 | XP_017919418.1 | 27 - 28 | 4067 | 98 | ||
| XM_018063931.1 | XP_017919420.1 | 27 - 28 | 4058 | 98 | ||
| XM_018063932.1 | XP_017919421.1 | 28 - 29 | 4169 | 98 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map