Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 2 - 3 | 549 | 95 | |
| XM_018063927.1 | XP_017919416.1 | 2 - 3 | 200 | 95 | ||
| XM_018063928.1 | XP_017919417.1 | 2 - 3 | 151 | 95 | ||
| XM_018063931.1 | XP_017919420.1 | 2 - 3 | 549 | 95 | ||
| XM_018063932.1 | XP_017919421.1 | 2 - 3 | 549 | 95 | ||
| GenBank mRNA | JO573590.2 | - | 2 - 3 | 207 | 95 | |
| KA327226.1 | - | 2 - 3 | 263 | 95 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map