Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 30 - 31 | 4413 | 122 | |
| XM_018063927.1 | XP_017919416.1 | 30 - 31 | 4064 | 122 | ||
| XM_018063928.1 | XP_017919417.1 | 30 - 31 | 4015 | 122 | ||
| XM_018063929.1 | XP_017919418.1 | 29 - 30 | 4311 | 122 | ||
| XM_018063931.1 | XP_017919420.1 | 29 - 30 | 4302 | 122 | ||
| XM_018063932.1 | XP_017919421.1 | 30 - 31 | 4413 | 122 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map