Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 33 - 34 | 4736 | 94 | |
| XM_018063927.1 | XP_017919416.1 | 33 - 34 | 4387 | 94 | ||
| XM_018063928.1 | XP_017919417.1 | 33 - 34 | 4338 | 94 | ||
| XM_018063929.1 | XP_017919418.1 | 32 - 33 | 4634 | 94 | ||
| XM_018063931.1 | XP_017919420.1 | 32 - 33 | 4625 | 94 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map