Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 34 - 35 | 4835 | 86 | |
| XM_018063927.1 | XP_017919416.1 | 34 - 35 | 4486 | 86 | ||
| XM_018063928.1 | XP_017919417.1 | 34 - 35 | 4437 | 86 | ||
| XM_018063929.1 | XP_017919418.1 | 33 - 34 | 4733 | 86 | ||
| XM_018063931.1 | XP_017919420.1 | 33 - 34 | 4724 | 86 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map