Assay Details
| Gene Symbol: LOC102178109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063926.1 | XP_017919415.1 | 36 - 37 | 5096 | 81 | |
| XM_018063927.1 | XP_017919416.1 | 36 - 37 | 4747 | 81 | ||
| XM_018063928.1 | XP_017919417.1 | 36 - 37 | 4698 | 81 | ||
| XM_018063929.1 | XP_017919418.1 | 35 - 36 | 4994 | 81 | ||
| XM_018063931.1 | XP_017919420.1 | 35 - 36 | 4985 | 81 | ||
More Information
Set Membership: |
|

LOC102178109
Genomic Map