Assay Details
| Gene Symbol: ATPSCKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972873.2 | XP_013828327.2 | 3 - 4 | 596 | 81 | |
| XM_018065549.1 | XP_017921038.1 | 3 - 4 | 529 | 81 | ||
| XM_018065550.1 | XP_017921039.1 | 3 - 4 | 529 | 81 | ||
| GenBank mRNA | JO371210.2 | - | 1 - 2 | 58 | 81 | |
More Information
Set Membership: |
|

ATPSCKMT
Genomic Map