Assay Details
| Gene Symbol: ZSCAN21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039892.1 | XP_017895381.1 | 2 - 3 | 585 | 72 | |
| XM_018039893.1 | XP_017895382.1 | 2 - 3 | 652 | 72 | ||
| XM_018039894.1 | XP_017895383.1 | 2 - 3 | 518 | 72 | ||
| GenBank mRNA | JO430440.2 | - | 3 - 4 | 598 | 72 | |
More Information
Set Membership: |
|

ZSCAN21
Genomic Map