Assay Details
| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041635.1 | XP_017897124.1 | 18 - 19 | 2281 | 62 | |
| XM_018041636.1 | XP_017897125.1 | 18 - 19 | 2275 | 62 | ||
| XM_018041637.1 | XP_017897126.1 | 18 - 19 | 2281 | 62 | ||
| XM_018041638.1 | XP_017897127.1 | 18 - 19 | 2281 | 62 | ||
| XM_018041639.1 | XP_017897128.1 | 18 - 19 | 2275 | 62 | ||
| XM_018041640.1 | XP_017897129.1 | 17 - 18 | 2059 | 62 | ||
More Information
Set Membership: |
|

JAKMIP3
Genomic Map