Assay Details
| Gene Symbol: LOC102179921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975998.2 | XP_013831452.2 | 2 - 3 | 566 | 66 | |
| GenBank mRNA | JO412362.2 | - | 2 - 3 | 573 | 66 | |
More Information
Set Membership: |
|

LOC102179921
Genomic Map