Assay Details
| Gene Symbol: LONRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044942.1 | XP_017900431.1 | 10 - 11 | 2287 | 55 | |
| XM_018044943.1 | XP_017900432.1 | 11 - 12 | 2591 | 55 | ||
More Information
Set Membership: |
|

LONRF3
Genomic Map