Assay Details
| Gene Symbol: STK26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044258.1 | XP_017899747.1 | 4 - 5 | 624 | 70 | |
| GenBank mRNA | JO577166.1 | - | 4 - 5 | 665 | 70 | |
More Information
Set Membership: |
|

STK26
Genomic Map