Assay Details
| Gene Symbol: DAXX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038577.1 | XP_017894066.1 | 6 - 7 | 2116 | 74 | |
| XM_018038578.1 | XP_017894067.1 | 6 - 7 | 2109 | 74 | ||
| GenBank mRNA | JT674559.1 | - | 2 - 3 | 437 | 74 | |
More Information
Set Membership: |
|

DAXX
Genomic Map