Assay Details
| Gene Symbol: CXCL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681789.3 | XP_005681846.2 | 3 - 4 | 346 | 134 | |
| XM_013964736.2 | XP_013820190.2 | 3 - 4 | 313 | 134 | ||
| GenBank mRNA | JO612593.2 | - | 3 - 4 | 376 | 134 | |
More Information
Set Membership: |
|

CXCL9
Genomic Map