Assay Details
| Gene Symbol: LOC102169972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687333.3 | XP_005687390.3 | 4 - 5 | 1393 | 63 | |
| XM_018056143.1 | XP_017911632.1 | 5 - 6 | 661 | 63 | ||
| XM_018056144.1 | XP_017911633.1 | 4 - 5 | 597 | 63 | ||
| GenBank mRNA | JO347079.2 | - | 3 - 4 | 455 | 63 | |
More Information
Set Membership: |
|

LOC102169972
Genomic Map