Assay Details
| Gene Symbol: TNFRSF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060552.1 | XP_017916041.1 | 3 - 4 | 644 | 63 | |
| XM_018060553.1 | XP_017916042.1 | 3 - 4 | 645 | 63 | ||
| GenBank mRNA | KA346035.1 | - | 1 - 2 | 727 | 63 | |
More Information
Set Membership: |
|

TNFRSF18
Genomic Map