Assay Details
| Gene Symbol: GOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692097.3 | XP_005692154.2 | 1 - 2 | 275 | 56 | |
| GenBank mRNA | JO360125.2 | - | 9 - 10 | 1563 | 56 | |
| JO576271.1 | - | 1 - 2 | 282 | 56 | ||
More Information
Set Membership: |
|

GOT2
Genomic Map