Assay Details
| Gene Symbol: SAP30BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694130.2 | XP_005694187.1 | 5 - 6 | 1020 | 62 | |
| XM_005694131.2 | XP_005694188.1 | 5 - 6 | 1020 | 62 | ||
| XM_005694133.3 | XP_005694190.1 | 2 - 3 | 177 | 62 | ||
| XM_013972463.2 | XP_013827917.1 | 5 - 6 | 449 | 62 | ||
| XM_013972464.2 | XP_013827918.1 | 5 - 6 | 1057 | 62 | ||
| GenBank mRNA | JO429423.2 | - | 5 - 6 | 430 | 62 | |
| KA304716.1 | - | 5 - 6 | 444 | 62 | ||
More Information
Set Membership: |
|

SAP30BP
Genomic Map