Assay Details
| Gene Symbol: ABHD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695892.3 | XP_005695949.2 | 5 - 6 | 964 | 69 | |
| XM_013973558.2 | XP_013829012.2 | 5 - 6 | 1115 | 69 | ||
| XM_018066914.1 | XP_017922403.1 | 5 - 6 | 1137 | 69 | ||
More Information
Set Membership: |
|

ABHD14A
Genomic Map