Assay Details
| Gene Symbol: LTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696604.3 | XP_005696661.1 | 3 - 4 | 1193 | 79 | |
| XM_018039178.1 | XP_017894667.1 | 2 - 3 | 1146 | 79 | ||
| GenBank mRNA | JO332344.2 | - | 1 - 2 | 148 | 79 | |
More Information
Set Membership: |
|

LTB
Genomic Map