Assay Details
| Gene Symbol: PI4K2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698386.3 | XP_005698443.3 | 6 - 7 | 1204 | 61 | |
| GenBank mRNA | JO427658.1 | - | 6 - 7 | 1000 | 61 | |
| JT691814.1 | - | 7 - 8 | 2547 | 61 | ||
More Information
Set Membership: |
|

PI4K2A
Genomic Map