Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698825.3 | XP_005698882.2 | 1 - 2 | 243 | 58 | |
| GenBank mRNA | KA319673.1 | - | 1 - 2 | 252 | 58 | |
More Information
Set Membership: |
|

BRF2
Genomic Map