Assay Details
| Gene Symbol: PCF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699475.3 | XP_005699532.1 | 10 - 11 | 4602 | 62 | |
| XM_005699476.3 | XP_005699533.1 | 10 - 11 | 4214 | 62 | ||
More Information
Set Membership: |
|

PCF11
Genomic Map