Assay Details
| Gene Symbol: LOC102190672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964448.2 | XP_013819902.2 | 1 - 2 | 582 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102190672| Gene Symbol: LOC102190672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964448.2 | XP_013819902.2 | 1 - 2 | 582 | 94 | |
Set Membership: |
|