Assay Details
Gene Symbol: SLC25A36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049798.1 | XP_017905287.1 | 4 - 5 | 657 | 70 | |
XM_018049811.1 | XP_017905300.1 | 4 - 5 | 665 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
