Assay Details
| Gene Symbol: LOC102187810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045395.1 | XP_017900884.1 | 16 - 17 | 2122 | 80 | |
| GenBank mRNA | JO427006.1 | - | 3 - 4 | 856 | 80 | |
| JO644629.2 | - | 3 - 4 | 800 | 80 | ||
More Information
Set Membership: |
|

LOC102187810
Genomic Map