Assay Details
| Gene Symbol: LOC102187810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045395.1 | XP_017900884.1 | 2 - 3 | 722 | 69 | |
| GenBank mRNA | JO427006.1 | - | 17 - 18 | 2256 | 69 | |
| JO560724.2 | - | 729 | 69 | |||
| JO644629.2 | - | 17 - 18 | 2200 | 69 | ||
More Information
Set Membership: |
|

LOC102187810
Genomic Map