Assay Details
| Gene Symbol: SRXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968785.2 | XP_013824239.1 | 1 - 2 | 478 | 82 | |
| GenBank mRNA | JO453227.2 | - | 1 - 2 | 288 | 82 | |
| JO589071.2 | - | 1 - 2 | 244 | 82 | ||
More Information
Set Membership: |
|

SRXN1
Genomic Map