Assay Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061554.1 | XP_017917043.1 | 8 - 9 | 1492 | 80 | |
XM_018061555.1 | XP_017917044.1 | 8 - 9 | 1305 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
