Assay Details
| Gene Symbol: PLBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061180.1 | XP_017916669.1 | 4 - 5 | 695 | 64 | |
| GenBank mRNA | JO632264.2 | - | 4 - 5 | 762 | 64 | |
More Information
Set Membership: |
|

PLBD2
Genomic Map