Assay Details
| Gene Symbol: HIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062420.1 | XP_017917909.1 | 1 - 2 | 1182 | 66 | |
| GenBank mRNA | JO435449.2 | - | 1 - 2 | 503 | 66 | |
More Information
Set Membership: |
|

HIPK4
Genomic Map