Assay Details
| Gene Symbol: HIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062420.1 | XP_017917909.1 | 2 - 3 | 1534 | 87 | |
| GenBank mRNA | JO435449.2 | - | 2 - 3 | 855 | 87 | |
More Information
Set Membership: |
|

HIPK4
Genomic Map