Assay Details
| Gene Symbol: LINGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066520.1 | XP_017922009.1 | 3 - 4 | 233 | 76 | |
| XM_018066524.1 | XP_017922013.1 | 3 - 4 | 234 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018066520.1, XM_018066524.1 transcripts.
Set Membership: |
|

LINGO1
Genomic Map