Assay Details
| Gene Symbol: PNPLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973931.2 | XP_013829385.1 | 7 - 8 | 1942 | 62 | |
| XM_018039266.1 | XP_017894755.1 | 7 - 8 | 1939 | 62 | ||
| XM_018039267.1 | XP_017894756.1 | 7 - 8 | 1943 | 62 | ||
| GenBank mRNA | KA327333.1 | - | 7 - 8 | 1980 | 62 | |
More Information
Set Membership: |
|

PNPLA1
Genomic Map