Assay Details
| Gene Symbol: TPGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974316.2 | XP_013829770.1 | 2 - 3 | 188 | 63 | |
| GenBank mRNA | JO434262.2 | - | 2 - 3 | 296 | 63 | |
More Information
Set Membership: |
|

TPGS2
Genomic Map