Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966023.2 | XP_013821477.1 | 3 - 4 | 357 | 73 | |
| XM_018049770.1 | XP_017905259.1 | 5 - 6 | 625 | 73 | ||
More Information
Set Membership: |
|

PXYLP1
Genomic Map